EMAIL THIS PAGE TO A FRIEND
223204 Aldrich

TES

Synonym: 2-[(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]ethanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid, TES free acid

  • CAS Number 7365-44-8

  • Empirical Formula (Hill Notation) C6H15NO6S

  • Molecular Weight 229.25

  •  Beilstein Registry Number 1957061

  •  EC Number 230-906-3

  •  MDL number MFCD00007532

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   JOCBASBOOFNAJA-UHFFFAOYSA-N
useful pH range   6.8 - 8.2
pKa (25 °C)   7.5
mp   ~225 °C (dec.)

Description

Application

Physiological buffer, pKa=7.5 at 20°C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?