• USA Home
  • 225215 - 2-Ethoxy-1-naphthoic acid

EMAIL THIS PAGE TO A FRIEND
225215 Aldrich

2-Ethoxy-1-naphthoic acid

97%

  • CAS Number 2224-00-2

  • Empirical Formula (Hill Notation) C13H12O3

  • Molecular Weight 216.23

  •  PubChem Substance ID 329752323

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   MYFBSSDLYGWAHH-UHFFFAOYSA-N
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?