EMAIL THIS PAGE TO A FRIEND
226556 Aldrich

Tridecanoic acid

technical grade, 90%

  • CAS Number 638-53-9

  • Linear Formula CH3(CH2)11CO2H

  • Molecular Weight 214.34

  •  Beilstein Registry Number 508317

  •  MDL number MFCD00002741

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   technical grade
InChI Key   SZHOJFHSIKHZHA-UHFFFAOYSA-N
assay   90%
bp   236 °C/100 mmHg(lit.)
mp   41-42 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?