EMAIL THIS PAGE TO A FRIEND
230456 Aldrich

Thioflavin S

Synonym: Direct Yellow 7, Thioflavine S

Purchase

Properties

absorption   λmax 374 nm

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T1892 Thioflavine S, practical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?