EMAIL THIS PAGE TO A FRIEND
23193 Aldrich

4-tert-Butylcatechol

technical, flakes, ≥98% (HPLC)

  • CAS Number 98-29-3

  • Linear Formula (CH3)3CC6H3-1,2-(OH)2

  • Molecular Weight 166.22

  •  Beilstein Registry Number 2043335

  •  EC Number 202-653-9

  •  MDL number MFCD00002201

  •  PubChem Substance ID 57648024

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   XESZUVZBAMCAEJ-UHFFFAOYSA-N
grade   technical
assay   ≥98% (HPLC)
quality   flakes
impurities   ≤1% 3,5-di-tert.-butylpyrocat. (HPLC)
bp   285 °C(lit.)
mp   50-56 °C
  52-55 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2923 8/PG 3
WGK Germany 
3
RTECS 
UX1400000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?