EMAIL THIS PAGE TO A FRIEND
232009 Aldrich

Nonyl phenyl ketone

98%

Synonym: Decanophenone

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   QQXJNLYVPPBERR-UHFFFAOYSA-N
assay   98%
bp   168 °C/5 mmHg(lit.)
mp   34-36 °C(lit.)

Description

Application

Nonyl phenyl ketone along with pyrene was employed as fluorescent probe-quencher to investigate the micelle size of polysoaps by luminescence quenching.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?