• USA Home
  • 233684 - Mercury(II) tetrathiocyanatocobaltate(II)

EMAIL THIS PAGE TO A FRIEND
233684 Aldrich

Mercury(II) tetrathiocyanatocobaltate(II)

Synonym: Cobalt(II) tetrathiocyanatomercurate(II), Mercury thiocyanatocobaltate(II)

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Materials Science, Mercury,
InChI Key   CKGHPFRRZBSWMC-UHFFFAOYSA-J
form   powder and chunks

Description

Application

High-purity magnetic-susceptibility standard

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2025 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?