EMAIL THIS PAGE TO A FRIEND

234400 Aldrich

Epiandrosterone

97%

Synonym: 3β-Hydroxy-5α-androstan-17-one, 3β-Hydroxyetioallocholan-17-one, 5α-Androstan-3β-ol-17-one, trans-Androsterone, Epi-androsterone, Isoandrosterone

  • CAS Number 481-29-8

  • Empirical Formula (Hill Notation) C19H30O2

  • Molecular Weight 290.44

  •  Beilstein Registry Number 1884007

  •  EC Number 207-563-3

  •  MDL number MFCD00064134

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

assay   97%
optical activity   [α]25/D +92°, c = 1 in methanol
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
mp   172-174 °C(lit.)

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Safety Statements (Europe) 
22-24/25
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Merck 14,3609

Beil. 8,IV,642

Corp MSDS 1, 1515:C / FT-IR 2 (3), 4225:A / FT-IR 1 (2), 1050:C / FT-NMR 1 (3), 573:A / NMR-Reference 2 (2), 921:D / RegBook 1 (2), 2857:L / Sigma FT-IR 1 (1), 1239:A / Structure Index 1, 457:C:8

Related Products

replaced by

Product #

Description

Add to Cart

E3375 trans-Androsterone

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?