• USA Home
  • 234427 - Benzalkonium chloride

EMAIL THIS PAGE TO A FRIEND
234427 Aldrich

Benzalkonium chloride

Synonym: Alkylbenzyldimethylammonium chloride, Alkyldimethylbenzylammonium chloride

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3261 - class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
WGK Germany 
2
RTECS 
BO3151000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?