• USA Home
  • 234516 - 2-Bromoethylamine hydrobromide

EMAIL THIS PAGE TO A FRIEND
234516 Aldrich

2-Bromoethylamine hydrobromide

95%

Synonym: 2-Aminoethyl bromide hydrobromide

  • CAS Number 2576-47-8

  • Linear Formula BrCH2CH2NH2 · HBr

  • Molecular Weight 204.89

  •  Beilstein Registry Number 3607605

  •  EC Number 219-924-2

  •  MDL number MFCD00012886

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   WJAXXWSZNSFVNG-UHFFFAOYSA-N
assay   95%
mp   170-175 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
KQ8225000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?