• USA Home
  • 234664 - (−)-Sparteine sulfate pentahydrate

EMAIL THIS PAGE TO A FRIEND
234664 Aldrich

(−)-Sparteine sulfate pentahydrate

99%

Synonym: (−)-Sparteine sulfate salt, Lupinidine sulfate pentahydrate

  • CAS Number 6160-12-9

  • Empirical Formula (Hill Notation) C15H26N2 · H2SO4 · 5H2O

  • Molecular Weight 422.54

  •  MDL number MFCD00149333

  •  PubChem Substance ID 24854051

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Deprotonation
InChI Key   WNSDDGBLIALDPB-LIELMIIZSA-N
assay   99%
optical activity   [α]25/D −12.4°, c = 1 in ethanol
mp   133-140 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?