• USA Home
  • 235423 - 3-Hydroxy-4-methoxymandelic acid

EMAIL THIS PAGE TO A FRIEND
235423 Aldrich

3-Hydroxy-4-methoxymandelic acid

98%

Synonym: Isovanillylmandelic acid

  • CAS Number 3695-24-7

  • Linear Formula HOC6H3(OCH3)CH(OH)CO2H

  • Molecular Weight 198.17

  •  Beilstein Registry Number 3202282

  •  EC Number 223-017-7

  •  MDL number MFCD00004230

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   PXMUSCHKJYFZFD-UHFFFAOYSA-N
assay   98%
mp   130-135 °C(lit.)

Description

Packaging

Packaged in glass bottles

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?