EMAIL THIS PAGE TO A FRIEND
236888 Aldrich

Benzene-d6

99.6 atom % D

Synonym: Hexadeuterobenzene

Purchase

Properties

Related Categories Alphabetical Listings, B, Benzene-d6, NMR Solvents, Solvent by Application,
InChI Key   UHOVQNZJYSORNB-MZWXYZOWSA-N
isotopic purity   99.6 atom % D
packaging   pk of 10 × 1 mL
refractive index   n20/D 1.497(lit.)
bp   79.1 °C/760 mmHg(lit.)
mp   6.8 °C(lit.)
density   0.950 g/mL at 25 °C(lit.)
mass shift   M+6

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1114 - class 3 - PG 2 - Benzene
WGK Germany 
3
Flash Point(F) 
12.2 °F
Flash Point(C) 
-11 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?