Synonym: Methyl-d3 alcohol d, Tetradeuteromethanol
CAS Number 811-98-3
Linear Formula CD3OD
Molecular Weight 36.07
Beilstein Registry Number 1733278
EC Number 212-378-6
MDL number MFCD00044637
PubChem Substance ID 24854205
Popular Documents: FTNMR (PDF)
| Related Categories | Aldrich 100% NMR Solvents for High Resolution, Alphabetical Listings, High Resolution NMR, High Throughput NMR, Labware, |
| isotopic purity | 99.96 atom % D |
| mol wt | mol wt 36.07 |
| packaging | pk of 10 × 0.5 mL |
| refractive index |
n |
| bp | 65.4 °C(lit.) |
| density | 0.888 g/mL at 25 °C(lit.) |
1 pkg in ampule
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?