EMAIL THIS PAGE TO A FRIEND
237027 Aldrich

Dichloromethane-d2

"100%", 99.96 atom % D

Synonym: Dideuteromethylenechloride, Methylene chloride-d2

Purchase

Properties

Related Categories Dichloromethane (Methylene chloride), Dichloromethane-d2, NMR Solvents, Solvent by Type, Solvent by Application,
vapor pressure   25.59 psi ( 55 °C)
  7.63 psi ( 20 °C)
InChI Key   YMWUJEATGCHHMB-DICFDUPASA-N
isotopic purity   99.96 atom % D
packaging   pk of 10 × 0.5 mL
refractive index   n20/D 1.422(lit.)
bp   40 °C(lit.)
density   1.362 g/mL at 25 °C(lit.)
mass shift   M+2

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?