• USA Home
  • 237779 - (S)-(+)-1,2-Propanediol

EMAIL THIS PAGE TO A FRIEND
237779 Aldrich

(S)-(+)-1,2-Propanediol

99%

Synonym: (S)-(+)-Propylene glycerol

  • CAS Number 4254-15-3

  • Linear Formula CH3CH(OH)CH2OH

  • Molecular Weight 76.09

  •  Beilstein Registry Number 1718871

  •  MDL number MFCD00004539

  •  PubChem Substance ID 24854270

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   DNIAPMSPPWPWGF-VKHMYHEASA-N
assay   99%
optical activity   [α]20/D +16.6°, neat
refractive index   n20/D 1.432(lit.)
bp   186-188 °C/765 mmHg(lit.)
density   1.036 g/mL at 20 °C(lit.)

Description

Application

Chiral synthetic intermediate and substrate for enzyme studies.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
Flash Point(F) 
217 °F
Flash Point(C) 
103 °C
Protocols & Articles

Articles

GC Analysis of 1,2-Alkyldiol Enantiomers (O-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of 1,2-Alkyldiol Enantiomers (O-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?