EMAIL THIS PAGE TO A FRIEND
237809 Aldrich

1-Triacontanol

96%

Synonym: 1-Hydroxytriacontane, Melissyl alcohol

Purchase

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T3777 1-Triacontanol, ≥98% (capillary GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?