• USA Home
  • 238791 - 2,4-Diethyl-1-heptanol

EMAIL THIS PAGE TO A FRIEND
238791 Aldrich

2,4-Diethyl-1-heptanol

95%

  • CAS Number 80192-55-8

  • Empirical Formula (Hill Notation) C11H24O

  • Molecular Weight 172.31

  •  PubChem Substance ID 329752512

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   MPSSVYBWYAOYHH-UHFFFAOYSA-N
assay   95%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?