• USA Home
  • 240044 - 2-(Diethylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
240044 Aldrich

2-(Diethylamino)ethanol

≥99%

Synonym: N,N-Diethylethanolamine, DEAE, DEEA

Purchase

Properties

vapor density   4.04 (vs air)
vapor pressure   1 mmHg ( 20 °C)
InChI Key   BFSVOASYOCHEOV-UHFFFAOYSA-N
assay   ≥99%
expl. lim.   11.7 %
refractive index   n20/D 1.441(lit.)
bp   161 °C(lit.)
density   0.884 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2686 8/PG 2
WGK Germany 
1
RTECS 
KK5075000
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?