• USA Home
  • 240397 - (±)-Propylene oxide

EMAIL THIS PAGE TO A FRIEND
240397 Aldrich

(±)-Propylene oxide

ReagentPlus®, ≥99%

Synonym: (±)-Methyloxirane, 1,2-Epoxypropane

  • CAS Number 75-56-9

  • Empirical Formula (Hill Notation) C3H6O

  • Molecular Weight 58.08

  •  Beilstein Registry Number 79763

  •  EC Number 200-879-2

  •  MDL number MFCD00005126

  •  PubChem Substance ID 24854416

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Epoxide Monomers, Materials Science, Monomers, Polymer Science
grade   ReagentPlus®
vapor density   2 (vs air)
vapor pressure   29.43 psi ( 55 °C)
  8.59 psi ( 20 °C)
InChI Key   GOOHAUXETOMSMM-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   1378 °F
expl. lim.   37 %
refractive index   n20/D 1.366(lit.)
bp   34 °C(lit.)
mp   −112 °C(lit.)
density   0.83 g/mL at 25 °C(lit.)

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1280 - class 3 - PG 1 - Propylene oxide
WGK Germany 
3
RTECS 
TZ2975000
Flash Point(F) 
-34.6 °F
Flash Point(C) 
-37 °C
Protocols & Articles

Related Content

Carcinogen Selector - Digestive System - Stomach

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Stomach
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Digestive System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Adrenal Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Adrenal Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Mammary Gland

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Mammary Gland
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Nasal Cavity

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Reproductive System - Nasal Cavity
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Respiratory System
Keywords: Cancer, Carcinogens, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?