• USA Home
  • 241113 - 2-Methylcyclohexanol, mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
241113 Aldrich

2-Methylcyclohexanol, mixture of cis and trans

≥99%

Synonym: Hexahydro-o-cresol

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   NDVWOBYBJYUSMF-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.462(lit.)
bp   163-166 °C(lit.)
density   0.93 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2617 3 / PGIII
WGK Germany 
1
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C
Protocols & Articles

Articles

GC Analysis of Industrial Solvents on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?