EMAIL THIS PAGE TO A FRIEND
241210 Aldrich

Mandelic acid

≥99%

Purchase

Properties

InChI Key   IWYDHOAUDWTVEP-UHFFFAOYSA-N
assay   ≥99%
mp   119-121 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
OO6300000
Protocols & Articles

Articles

GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?