• USA Home
  • 241504 - Filling solution for salt bridges and double-junction electrodes

EMAIL THIS PAGE TO A FRIEND
241504 Aldrich

Filling solution for salt bridges and double-junction electrodes

17% aqueous KNO3

Purchase

Properties

Related Categories Analytical/Chromatography, Electrolytes, Ion Sensor Materials More...
concentration   17% aqueous KNO3
density   1.055 g/mL at 25 °C

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?