EMAIL THIS PAGE TO A FRIEND
243337 Aldrich

Filter agent

Celatom® FW-60

Purchase

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Diatomaceous Earth, Essential Chemicals, Research Essentials
InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N
parameter   3000 flow rate
  5.000 (Permeability(Darcy))
color   936

Description

Legal Information

Celatom is a registered trademark of EP Minerals, LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?