• USA Home
  • 246891 - Chlorotribenzylsilane

EMAIL THIS PAGE TO A FRIEND
246891 Aldrich

Chlorotribenzylsilane

97%

Synonym: Tribenzylchlorosilane, Tribenzylsilyl chloride

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
InChI Key   UTXPCJHKADAFBB-UHFFFAOYSA-N
assay   97%
mp   139-142 °C(lit.)

Description

Application

Reagent for OH protection.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?