• USA Home
  • 247235 - 4,6-Dimethyl-2-nitroaniline

EMAIL THIS PAGE TO A FRIEND
247235 Aldrich

4,6-Dimethyl-2-nitroaniline

technical grade, 90%

Purchase

Properties

Related Categories Amines, Building Blocks, C8, Chemical Synthesis, Nitrogen Compounds,
grade   technical grade
InChI Key   VSRYYONYIUUFFY-UHFFFAOYSA-N
assay   90%
mp   64-68 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?