• USA Home
  • 247928 - Selenium atomic absorption standard solution

EMAIL THIS PAGE TO A FRIEND
247928 Aldrich

Selenium atomic absorption standard solution

1,000 μg/mL Se in 1 wt. % HNO3 (Exact Se concentration on label)

Purchase

Properties

concentration   1,000 μg/mL Se in 1 wt. % HNO3 (Exact Se concentration on label)
density   1.006 g/mL at 25 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3264 8/PG 3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

89498 Selenium Standard for AAS, TraceCERT®, 1000 mg/L Se in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?