• USA Home
  • 248096 - 2,3,4,5,6-Pentafluorophenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
248096 Aldrich

2,3,4,5,6-Pentafluorophenylacetonitrile

99%

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C7-C8, C8 to C9, Chemical Synthesis,
InChI Key   YDNOJUAQBFXZCR-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.439(lit.)
bp   107-111 °C/17 mmHg(lit.)
density   1.61 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?