• USA Home
  • 249203 - trans-7, cis-9-Dodecadienyl acetate

EMAIL THIS PAGE TO A FRIEND
249203 Aldrich

trans-7, cis-9-Dodecadienyl acetate

98%

Synonym: (E,Z)-7,9-DDDA

Purchase

Properties

InChI Key   LLRZUAWETKPZJO-SCFJQAPRSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
141.8 °F
Flash Point(C) 
61 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?