• USA Home
  • 249653 - 2-Acetyl-4-methylthiophene

EMAIL THIS PAGE TO A FRIEND
249653 Aldrich

2-Acetyl-4-methylthiophene

97%

Synonym: 1-(4-Methyl-2-thienyl)ethanone

  • CAS Number 13679-73-7

  • Empirical Formula (Hill Notation) C7H8OS

  • Molecular Weight 140.20

  •  MDL number MFCD00027395

Purchase

Properties

InChI Key   NFZNHUNPIJPRMP-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.5600(lit.)
bp   108-109 °C(lit.)
density   1.224 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?