• USA Home
  • 25112 - (R)-2-Chloro-3-methylbutyric acid

EMAIL THIS PAGE TO A FRIEND
25112 Aldrich

(R)-2-Chloro-3-methylbutyric acid

technical, ≥90% (GC)

Synonym: (R)-α-Chloroisovaleric acid

  • CAS Number 84918-96-7

  • Empirical Formula (Hill Notation) C5H9ClO2

  • Molecular Weight 136.58

  •  Beilstein Registry Number 7461753

  •  MDL number MFCD00044962

  •  PubChem Substance ID 24855106

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   DDTJFSPKEIAZAM-SCSAIBSYSA-N
grade   technical
assay   ≥90% (GC)
optical activity   [α]20/D 0.0 to +1.5°, c = neat
refractive index   n20/D 1.443
density   1.144 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?