• USA Home
  • 251704 - 3-(Amidinothio)-1-propanesulfonic acid

EMAIL THIS PAGE TO A FRIEND
251704 Aldrich

3-(Amidinothio)-1-propanesulfonic acid

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
InChI Key   MQLJIOAPXLAGAP-UHFFFAOYSA-N
assay   97%
mp   254-256 °C(lit.)
solubility   water: soluble25 mg/mL, clear, colorless

Description

Application

3-(Amidinothio)-1-propanesulfonic acid may be used in chemical synthesis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?