EMAIL THIS PAGE TO A FRIEND
251925 Aldrich

(−)-Borneol

99%

Synonym: endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

  • CAS Number 464-45-9

  • Empirical Formula (Hill Notation) C10H18O

  • Molecular Weight 154.25

  •  Beilstein Registry Number 2038053

  •  EC Number 207-353-1

  •  MDL number MFCD00003759

  •  PubChem Substance ID 24855169

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
vapor density   5.31 (vs air)
vapor pressure   33.5 mmHg ( 25 °C)
InChI Key   DTGKSKDOIYIVQL-QXFUBDJGSA-N
assay   99%
optical activity   [α]20/D −35.6°, c = 5 in ethanol
bp   210 °C(lit.)
mp   206-208 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1312 - class 4.1 - PG 3 - Borneol
WGK Germany 
2
RTECS 
DT5095000
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

139114 (−)-Borneol, predominantly endo, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?