• USA Home
  • 253723 - o-Tolyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
253723 Aldrich

o-Tolyl isothiocyanate

99%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocyanates/Isothiocyanates More...
InChI Key   JYKYYPPZLPVIBY-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.6361(lit.)
bp   239 °C(lit.)
density   1.115 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
Flash Point(F) 
226.4 °F
Flash Point(C) 
108 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?