Skip to Content
MilliporeSigma

254592

2,4(3H,5H)-Furandione

98%

Synonym(s):

β-Tetronic acid, 4-Hydroxy-2(5H)-furanone, Tetrahydrofuran-2,4-dione, Tetronic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C4H4O3
CAS Number:
Molecular Weight:
100.07
Beilstein/REAXYS Number:
108442
EC Number:
MDL number:
UNSPSC Code:
12352103

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

98%

mp

141-143 °C (dec.) (lit.)

SMILES string

O=C1COC(=O)C1

Looking for similar products? Visit Product Comparison Guide

Application

Strongly acidic building block for heterocyclic systems[1] including medicinal intermediates.[2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liebigs Ann. Chem., 69-69 (1988)
Chemical & Pharmaceutical Bulletin, 32, 329-329 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service