• USA Home
  • 257230 - Trimethylaluminum solution

EMAIL THIS PAGE TO A FRIEND
257230 Aldrich

Trimethylaluminum solution

2.0 M in toluene

Purchase

Properties

Related Categories Chemical Synthesis, Organoaluminum, Organometallic Reagents More...
InChI Key   JLTRXTDYQLMHGR-UHFFFAOYSA-N
concentration   2.0 M in toluene
density   0.81 g/mL at 25 °C

Description

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3399A 4.3(3) / PGI
WGK Germany 
2
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

septum inlet adapter

Product #

Description

Add to Cart

Z118206 Angled septum-inlet adapter, brass, inlet I.D. 6 mm × 6 mm
Z118141 Straight septum-inlet adapter, inlet I.D. 6 mm, brass
Z118192 Straight septum-inlet adapter, inlet I.D. 13.9 mm, brass

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?