• USA Home
  • 258245 - Biliverdin dihydrochloride

EMAIL THIS PAGE TO A FRIEND
258245 Aldrich

Biliverdin dihydrochloride

technical grade, 80%

  • CAS Number 55482-27-4

  • Empirical Formula (Hill Notation) C33H34N4O6 · 2HCl

  • Molecular Weight 655.57

  •  Beilstein Registry Number 6045097

Purchase

Properties

grade   technical grade
InChI Key   KTRFJVRKVQROKS-UXNIKXASSA-N
assay   80%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?