• USA Home
  • 259845 - 2,5-Bis(4-diethylaminophenyl)-1,3,4-oxadiazole

EMAIL THIS PAGE TO A FRIEND
259845 Aldrich

2,5-Bis(4-diethylaminophenyl)-1,3,4-oxadiazole

98%

Purchase

Properties

InChI Key   UZGVMZRBRRYLIP-UHFFFAOYSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?