• USA Home
  • 259934 - Sodium 2,4-dinitrobenzenesulfonate

EMAIL THIS PAGE TO A FRIEND
259934 Aldrich

Sodium 2,4-dinitrobenzenesulfonate

98%

Synonym: 2,4-Dinitrobenzenesulfonic acid sodium salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
vapor density   9.3 (vs air)
InChI Key   GSBYVRKLPCSLNV-UHFFFAOYSA-M
assay   98%
mp   >350 °C(lit.)

Description

Packaging

5, 25 g in glass bottle

Application

Sodium 2,4-dinitrobenzenesulfonate has been used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?