• USA Home
  • 260584 - 4-Hydroxybenzoylformic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
260584 Aldrich

4-Hydroxybenzoylformic acid sodium salt

97%

Synonym: 4-Hydroxyphenylglyoxylic acid sodium salt

  • CAS Number 54537-30-3

  • Empirical Formula (Hill Notation) C8H5NaO4

  • Molecular Weight 188.11

  •  EC Number 259-205-0

Purchase

Properties

InChI Key   SIZKOKCUGWOJMH-UHFFFAOYSA-M
assay   97%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?