EMAIL THIS PAGE TO A FRIEND
260746 Aldrich

Digitonin

Synonym: Digitin

  • CAS Number 11024-24-1

  • Empirical Formula (Hill Notation) C56H92O29

  • Molecular Weight 1229.31

  •  Beilstein Registry Number 78654

  •  EC Number 234-255-6

  •  MDL number MFCD00077729

Purchase

Properties

InChI Key   UVYVLBIGDKGWPX-KUAJCENISA-N
optical activity   [α]20/D −50.1°, c = 1 in acetic acid: water (3:1)
mol wt   micellar avg mol wt 70,000
aggregation number   60
CMC   <0.5 mM(20-25°C)
mp   230-240 °C (dec.)(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGIII
WGK Germany 
3
RTECS 
IH2050050
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D5628 Digitonin, ~50% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?