EMAIL THIS PAGE TO A FRIEND
260878 Aldrich

Phenanthrene

zone-refined, ≥99.5%

  • CAS Number 85-01-8

  • Empirical Formula (Hill Notation) C14H10

  • Molecular Weight 178.23

  •  Beilstein Registry Number 1905428

  •  EC Number 201-581-5

  •  MDL number MFCD00001168

  •  PubChem Substance ID 24855775

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

grade   zone-refined
InChI Key   YNPNZTXNASCQKK-UHFFFAOYSA-N
assay   ≥99.5%
bp   340 °C(lit.)
mp   98-100 °C(lit.)
density   1.063 g/mL at 25 °C(lit.)
Gene Information   human ... CYP1A2(1544)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?