EMAIL THIS PAGE TO A FRIEND
261122 Aldrich

Holmium

ingot, 99.9% trace metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Holmium, Materials Science, Metal and Ceramic Science,
InChI Key   KJZYNXUDTRRSPN-UHFFFAOYSA-N
assay   99.9% trace metals basis
form   ingot
density   8.8 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

457957 Holmium, chips, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?