• USA Home
  • 26224 - 4-(2-Chloropropionyl)phenylacetic acid

EMAIL THIS PAGE TO A FRIEND
26224 Aldrich

4-(2-Chloropropionyl)phenylacetic acid

≥98.0%

  • CAS Number 84098-73-7

  • Empirical Formula (Hill Notation) C11H11ClO3

  • Molecular Weight 226.66

  •  Beilstein Registry Number 3284980

  •  MDL number MFCD01862983

  •  PubChem Substance ID 57648164

Purchase

Properties

Related Categories Bifunctional Linkers, Chemical Biology, Chemical Synthesis, Linkers and Crosslinkers
InChI Key   SYKQUPRADSJCQY-UHFFFAOYSA-N
assay   ≥98.0%
mp   141-145 °C

Description

Other Notes

Photolabile linker used in solid-phase peptide synthesis

Packaging

1 g in poly tube

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?