• USA Home
  • 26255 - (3-Chloropropyl)tris(trimethylsiloxy)silane

EMAIL THIS PAGE TO A FRIEND
26255 Aldrich

(3-Chloropropyl)tris(trimethylsiloxy)silane

≥97.0%

  • CAS Number 18077-31-1

  • Empirical Formula (Hill Notation) C12H33ClO3Si4

  • Molecular Weight 373.18

  •  Beilstein Registry Number 1957472

  •  EC Number 241-985-9

  •  MDL number MFCD00054781

  •  PubChem Substance ID 24855750

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Trialkoxysilanes
InChI Key   MMWCHSBIWFYBBL-UHFFFAOYSA-N
assay   ≥97.0%
refractive index   n20/D 1.413
bp   181 °C/100 mmHg(lit.)
density   0.927 g/mL at 20 °C(lit.)

Description

Packaging

5 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?