EMAIL THIS PAGE TO A FRIEND
263206 Aldrich

Terbium

chips, 99.9% trace rare earth metals basis

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science, Metals,
InChI Key   GZCRRIHWUXGPOV-UHFFFAOYSA-N
assay   99.9% trace rare earth metals basis
form   chips
resistivity   116 μΩ-cm, 20°C
bp   3230 °C(lit.)
mp   1356 °C(lit.)
density   8.234 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Functional Magnetic Materials: Synergy Between Basic Science and Evolving Technology

S. B. Roy Materials and Advanced Accelerator Sciences Division Raja Ramanna Centre for Advanced Technology Indore-452013, India Email: sbroy@rrcat.gov.in
Keywords: Crystallization, Microscopy, Phase transitions, Reductions, transformation

Kinetic Energy Harvesting by Magnetostrictive Materials

Daniele Davino Department of Engineering University of Sannio, 82100 Benevento, Italy Email: davino@unisannio.it
Keywords: Electronics, Environmental

Metal Hydrides for NiMH Battery Applications

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications. The Partnership for a New Generation of V...
Dhanesh Chandra,* Wen-Ming Chien and Anjali Talekar
Material Matters Volume 6 Article 2
Keywords: Absorption, Applications, Automotive, Capillary electrophoresis, Combustion, Degradations, Electrochemical analysis, Electronics, Environmental, Help, Ion-exchange chromatography, Phase transitions, Precipitation, Purification, Separation, Substitutions, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?