EMAIL THIS PAGE TO A FRIEND
265489 Aldrich

Niobium

rod, diam. 12.7 mm, ≥99.9% trace metals basis

Synonym: Columbium

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Metals, Niobium
InChI Key   GUCVJGMIXFAOAE-UHFFFAOYSA-N
assay   ≥99.9% trace metals basis
form   rod
resistivity   13-16 μΩ-cm, 20°C
diam.   12.7 mm
bp   4742 °C(lit.)
mp   2468 °C(lit.)
density   8.57 g/mL at 25 °C(lit.)

Description

Quantity

10 g = 1 cm

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?