• USA Home
  • 266442 - Ethylene glycol bisthioglycolate

EMAIL THIS PAGE TO A FRIEND
266442 Aldrich

Ethylene glycol bisthioglycolate

96%

Synonym: Ethylene glycol bis(mercaptoacetate), Ethylene glycol bismercaptoacetate

  • CAS Number 123-81-9

  • Linear Formula (HSCH2CO2CH2-)2

  • Molecular Weight 210.27

  •  MDL number MFCD00004875

Purchase

Properties

InChI Key   PSYGHMBJXWRQFD-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.522(lit.)
bp   137-139 °C/2 mmHg(lit.)
density   1.313 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?