• USA Home
  • 266485 - (4-Ethoxybenzyl)triphenylphosphonium bromide

EMAIL THIS PAGE TO A FRIEND
266485 Aldrich

(4-Ethoxybenzyl)triphenylphosphonium bromide

99%

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Olefination, Synthetic Reagents, Wittig Reagents More...
InChI Key   MZBKKJRCQNVENM-UHFFFAOYSA-M
assay   99%
mp   214-215 °C(lit.)

Description

Application

Reactant for:
• Preparation of styrylquinolones by Wittig reaction
• Preparation of indolone-N-oxides as antiplasmodial and anticancer agents
• Assembly of pyrimidinone core of HIV integrase inhibitor raltegravir potassium, via Wittig reaction

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?