EMAIL THIS PAGE TO A FRIEND
266760 Aldrich

Hafnium

sponge, +20 mesh, 99.5% trace metals basis (purity excludes ~2% zirconium)

Purchase

Properties

Related Categories Hafnium, Materials Science, Metal and Ceramic Science, Metals
InChI Key   VBJZVLUMGGDVMO-UHFFFAOYSA-N
assay   99.5% trace metals basis (purity excludes ~2% zirconium)
form   sponge
resistivity   29.6 μΩ-cm, 0°C
particle size   +20 mesh
bp   4602 °C(lit.)
mp   2227 °C(lit.)
density   13.3 g/cm3(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3178 4.1 / PGIII
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?